missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Proteasome subunit beta type-3 Protein

A DNA sequence encoding the Homo sapiens (Human) Proteasome subunit beta type-3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6545-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16054060

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_002786.2
Proteasome subunit beta type-3 Protein
Research Use Only
PSMB3
Recombinant Protein
E. coli
SIMSYNGGAV MAMKGKNCVA IAADRRFGIQ AQMVTTDFQK IFPMGDRLYI GLAGLATDVQ TVAQRLKFRL NLYELKEGRQ IKPYTLMSMV ANLLYEKRFG PYYTEPVIAG LDPKTFKPFI CSLDLIGCPM VTDDFVVSGT CAEQMYGMCE SLWEPNMDPD HLFETISQAM LNAVDRDAVS GMGVIVHIIE KDKITTRTLK ARMD
Greater than 80% as determined by SDS-PAGE.
5691
500 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers