missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Proteasome subunit beta type-2 Protein
A DNA sequence encoding the Homo sapiens (Human) Proteasome subunit beta type-2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8364-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002785.1 | |
Proteasome subunit beta type-2 Protein | |
Research Use Only | |
PSMB2 | |
Recombinant Protein | |
E. coli | |
MEYLIGIQGP DYVLVASDRV AASNIVQMKD DHDKMFKMSE KILLLCVGEA GDTVQFAEYI QKNVQLYKMR NGYELSPTAA ANFTRRNLAD CLRSRTPYHV NLLLAGYDEH EGPALYYMDY LAALAKAPFA AHGYGAFLTL SILDRYYTPT ISRERAVELL RKCLEELQKR FILNLPTFSV RIIDKNGIHD LDNISFPKQG S | |
Greater than 90% as determined by SDS-PAGE. |
5690 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |