missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PROS1 / Protein

A DNA sequence encoding the Homo sapiens (Human) PROS1 /, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6533-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16023990

  • 463.00 EUR / 100µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_000304.2
PROS1 / Protein
Research Use Only
PROS1
Recombinant Protein
E. coli
ANSLLEETKQ GNLERECIEE LCNKEEAREV FENDPETDYF YPKYLVCLRS FQTGLFTAAR QSTNAYPDLR SCVNAIPDQC SPLPCNEDGY MSCKDGKASF TCTCKPGWQG EKCEFDINEC KDPSNINGGC SQICDNTPGS YHCSCKNGFV MLSNKKDCKD VDECSLKPSI CGTAVCKNIP GDFECECPEG YRYNLKSKSC EDIDECSENM CAQLCVNYPG GYTCYCDGKK GFKLAQDQKS CEVVSVCLPL NLDTKYELLY LAEQFAGVVL YLKFRLPEIS RFSAEFDFRT YDSEGVILYA ESIDHSAWLL IALRGGKIEV QLKNEHTSKI TTGGDVINNG LWNMVSVEEL EHSISIKIAK EAVMDINKPG PLFKPENGLL ETKVYFAGFP RKVESELIKP INPRLDGCIR SWNLMKQGAS GIKEIIQEKQ NKHCLVTVEK GSYYPGSGIA QFHIDYNNVS SAEGWHVNVT LNIRPSTGTG VMLALVSGNN TVPFAVSLVD STSEKSQDIL LSVENTVIYR IQALSLCSDQ QSHLEFRVNR NNLELSTPLK IETISHEDLQ RQLAVLDKAM KAKVATYLGG LPDVPFSATP VNAFYNGCME VNINGVQLDL DEAISKHNDI RAHSCPSVWK KTKNS
Greater than 90% as determined by SDS-PAGE.
5627
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers