missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Prohibitin Protein
A DNA sequence encoding the Homo sapiens (Human) Prohibitin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6488-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001268425.1 | |
Prohibitin Protein | |
Research Use Only | |
PHB | |
Recombinant Protein | |
E. coli | |
MAAKVFESIG KFGLALAVAG GVVNSALYNV DAGHRAVIFD RFRGVQDIVV GEGTHFLIPW VQKPIIFDCR SRPRNVPVIT GSKDLQNVNI TLRILFRPVA SQLPRIFTSI GEDYDERVLP SITTEILKSV VARFDAGELI TQRELVSRQV SDDLTERAAT FGLILDDVSL THLTFGKEFT EAVEAKQVAQ QEAERARFVV EKAEQQKKAA IISAEGDSKA AELIANSLAT AGDGLIELRK LEAAEDIAYQ LSRSRNITYL PAGQSVLLQL PQ | |
Greater than 90% as determined by SDS-PAGE. |
5245 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |