missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Programmed cell death protein 1

A DNA sequence encoding the Homo sapiens (Human) Programmed cell death protein 5, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6475-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16023570

  • 463.00 EUR / 100µg
Estimated Shipment: 10-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_004699.1
Programmed cell death protein 1
Research Use Only
PDCD5
Recombinant Protein
E. coli
ADEELEALRR QRLAELQAKH GDPGDAAQQE AKHREAEMRN SILAQVLDQS ARARLSNLAL VKPEKTKAVE NYLIQMARYG QLSEKVSEQG LIEILKKVSQ QTEKTTTVKF NRRKVMDSDE DDDY
Greater than 80% as determined by SDS-PAGE.
9141
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers