missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Prealbumin / Transthyretin / TTR / PALB Protein
A DNA sequence encoding the Homo sapiens (Human) Prealbumin / Transthyretin / TTR / PALB, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6848-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000362.1 | |
Prealbumin / Transthyretin / TTR / PALB Protein | |
Research Use Only | |
TTR | |
Recombinant Protein | |
E. coli | |
GPTGTGESKC PLMVKVLDAV RGSPAINVAV HVFRKAADDT WEPFASGKTS ESGELHGLTT EEEFVEGIYK VEIDTKSYWK ALGISPFHEH AEVVFTANDS GPRRYTIAAL LSPYSYSTTA VVTNPKE | |
Greater than 90% as determined by SDS-PAGE. |
7276 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |