missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human POU domain, class 5, transcription factor 1 Protein
A DNA sequence encoding the Homo sapiens (Human) POU domain, class 5, transcription factor 1, was expressed in the hosts and tags indicated.
160.00 EUR - 2090.00 EUR
Specifications
Accession Number | NP_001272916.1 |
---|---|
Gene ID (Entrez) | 5460 |
Name | POU domain, class 5, transcription factor 1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16003860
|
enQuireBio™
QP6517-EC-100UG |
100 μg |
463.00 EUR
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16023860
|
enQuireBio™
QP6517-EC-1MG |
1 mg |
2090.00 EUR
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16013860
|
enQuireBio™
QP6517-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16033860
|
enQuireBio™
QP6517-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16043860
|
enQuireBio™
QP6517-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16053860
|
enQuireBio™
QP6517-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001272916.1 | |
POU domain, class 5, transcription factor 1 Protein | |
Not determined. | |
Full Length | |
Human | |
His-B2M | |
Tris-based buffer, 50% glycerol |
5460 | |
Research Use Only | |
POU5F1 | |
Recombinant Protein | |
E. coli | |
MAGHLASDFA FSPPPGGGGD GPGGPEPGWV DPRTWLSFQG PPGGPGIGPG VGPGSEVWGI PPCPPPYEFC GGMAYCGPQV GVGLVPQGGL ETSQPEGEAG VGVESNSDGA SPEPCTVTPG AVKLEKEKLE QNPEESQDIK ALQKELEQFA KLLKQKRITL GYTQADVGLT LGVLFGKVFS QTTICRFEAL QLSFKNMCKL RPLLQKWVEE ADNNENLQEI CKAETLVQAR KRKRTSIENR VRGNLENLFL QCPKPTLQQI SHIAQQLGLE KDVVRVWFCN RRQKGKRSSS DYAQREDFEA AGSPFSGGPV SFPLAPGPHF GTPGYGSPHF TALYSSVPFP EGEAFPPVSV TTLGSPMHSN | |
Greater than 80% as determined by SDS-PAGE. |