missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human POLR3K Protein
A DNA sequence encoding the Homo sapiens (Human) POLR3K, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6512-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_057394.3 | |
POLR3K Protein | |
Research Use Only | |
POLR3K | |
Recombinant Protein | |
E. coli | |
MLLFCPGCGN GLIVEEGQRC HRFSCNTCPY VHNITRKVTN RKYPKLKEVD DVLGGAAAWE NVDSTAESCP KCEHPRAYFM QLQTRSADEP MTTFYKCCNA QCGHRWRD | |
Greater than 90% as determined by SDS-PAGE. |
51728 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |