missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Plexin-A1 Protein

A DNA sequence encoding the Homo sapiens (Human) Plexin-A1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8791-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16006491

  • 1525.00 EUR / 500µg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_115618.3
Plexin-A1 Protein
Research Use Only
PLXNA1
Recombinant Protein
Yeast
LNAGSDVAVS VGGRPCSFSW RNSREIRCLT PPGQSPGSAP IIININRAQL TNPEVKYNYT EDPTILRIDP EWSINSGGTL LTVTGTNLAT VREPRIRAKY GGIERENGCL VYNDTTMVCR APSVANPVRS PPELGERPDE LGFVMDNVRS LLVLNSTSFL YYPDPVL
Greater than 90% as determined by SDS-PAGE.
5361
500 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers