missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PLCZ1 Protein
A DNA sequence encoding the Homo sapiens (Human) PLCZ1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7781-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001317698.1 | |
PLCZ1 Protein | |
Research Use Only | |
PLCZ1 | |
Recombinant Protein | |
E. coli | |
MEMRWFLSKI QDDFRGGKIN LEKTQRLLEK LDIRCSYIHV KQIFKDNDRL KQGRITIEEF RAIYRIITHR EEIIEIFNTY SENRKILLAS NLAQFLTQEQ YAAEMSKAIA FEIIQKYEPI EEVRKAHQMS LEGFTRYMDS RECLLFKNEC RKVYQDMTHP LNDYFISSSH NTYLVSDQLL GPSDLWGYVS ALVKGCRCLE IDCWDGAQNE PVVYHGYTLT SKLLFKTVIQ AIHKYAFMTS DYPVVLSLEN HCSTAQQEVM ADNLQATFGE SLLSDMLDDF PDTLPSPEAL KFKILVKNKK IGTLKETHER KGSDKRGDNQ DKETGVKKLP GVMLFKKKKT RKLKIALALS DLVIYTKAEK FKSFQHSRLY QQFNENNSIG ETQARKLSKL RVHEFIFHTR KFITRIYPKA TRADS | |
Greater than 90% as determined by SDS-PAGE. |
89869 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |