missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Platelet glycoprotein Ib alpha chain Protein

A DNA sequence encoding the Homo sapiens (Human) Platelet glycoprotein Ib alpha chain, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6109-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16030860

  • 2090.00 EUR / 1mg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_000164.5
Platelet glycoprotein Ib alpha chain Protein
Research Use Only
GP1BA
Recombinant Protein
E. coli
HPICEVSKVA SHLEVNCDKR NLTALPPDLP KDTTILHLSE NLLYTFSLAT LMPYTRLTQL NLDRCELTKL QVDGTLPVLG TLDLSHNQLQ SLPLLGQTLP ALTVLDVSFN RLTSLPLGAL RGLGELQELY LKGNELKTLP PGLLTPTPKL EKLSLANNNL TELPAGLLNG LENLDTLLLQ ENSLYTIPKG FFGSHLLPFA FLHGNPWLCN CEILYFRRWL QDNAENVYVW KQGVDVKAMT SNVASVQCDN SDKFPVYKYP GKGCPTLGDE GDTDLYDYYP EEDTEGDKVR ATRTVVKFPT KAHTTPWGLF YSWSTASLDS QMPSSLHPTQ ESTKEQTTFP PRWTPNFTLH MESITFSKTP KSTTEPTPSP TTSEPVPEPA PNMTTLEPTP SPTTPEPTSE PAPSPTTPEP TPIPTIATSP TILVSATSLI TPKSTFLTTT KPVSLLESTK KTIPELDQPP KLRGVLQGHL ESSRNDPFLH PDFCCLLPL
Greater than 90% as determined by SDS-PAGE.
2811
1 mg
Not determined.
Extracellular Domain
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers