missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Placental Lactogen / CSH1 Protein
A DNA sequence encoding the Homo sapiens (Human) Placental Lactogen / CSH1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8713-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001308.1 | |
Placental Lactogen / CSH1 Protein | |
Research Use Only | |
CSH1 | |
Recombinant Protein | |
E. coli | |
VQTVPLSRLF DHAMLQAHRA HQLAIDTYQE FEETYIPKDQ KYSFLHDSQT SFCFSDSIPT PSNMEETQQK SNLELLRISL LLIESWLEPV RFLRSMFANN LVYDTSDSDD YHLLKDLEEG IQTLMGRLED GSRRTGQILK QTYSKFDTNS HNHDALLKNY GLLYCFRKDM DKVETFLRMV QCRSVEGSCG F | |
Greater than 90% as determined by SDS-PAGE. |
1442 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |