missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PLA2G2D / Phospholipase A2 IID Protein

A DNA sequence encoding the Homo sapiens (Human) PLA2G2D / Phospholipase A2 IID, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8240-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16083491

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001258743.1
PLA2G2D / Phospholipase A2 IID Protein
Research Use Only
PLA2G2D
Recombinant Protein
E. coli
ILNLNKMVKQ VTGKMPILSY WPYGCHCGLG GRGQPKDATD WCCQTHDCCY DHLKTQGCSI YKDYYRYNFS QGNIHCSDKG SWCEQQLCAC DKEVAFCLKR NLDTYQKRLR FYWRPHCRGQ TPGC
Greater than 90% as determined by SDS-PAGE.
26279
500 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers