missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Phospholipid scramblase 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Phospholipid scramblase 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6506-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16043780

  • 2090.00 EUR / 1mg
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_066928.1
Phospholipid scramblase 1 Protein
Research Use Only
PLSCR1
Recombinant Protein
E. coli
ESTGSQEQKS GVW
Greater than 80% as determined by SDS-PAGE.
5359
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers