missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Peroxisomal multifunctional enzyme type 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Peroxisomal multifunctional enzyme type 2, was expressed in the hosts and tags indicated.
202.00 EUR - 2155.00 EUR
Specifications
Accession Number | NP_000405.1 |
---|---|
Gene ID (Entrez) | 3295 |
Name | Peroxisomal multifunctional enzyme type 2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16017811
|
enQuireBio™
QP9186-YE-100UG |
100 μg |
565.00 EUR
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16057811
|
enQuireBio™
QP9186-YE-500UG |
500 μg |
1525.00 EUR
500µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16027811
|
enQuireBio™
QP9186-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16047811
|
enQuireBio™
QP9186-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16067811
|
enQuireBio™
QP9186-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16037811
|
enQuireBio™
QP9186-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000405.1 | |
Peroxisomal multifunctional enzyme type 2 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
3295 | |
Research Use Only | |
HSD17B4 | |
Recombinant Protein | |
Yeast | |
MGSPLRFDGR VVLVTGAGAG LGRAYALAFA ERGALVVVND LGGDFKGVGK GSLAADKVVE EIRRRGGKAV ANYDSVEEGE KVVKTALDAF GRIDVVVNNA GILRDRSFAR ISDEDWDIIH RVHLRGSFQV TRAAWEHMKK QKYGRIIMTS SASGIYGNFG QANYSAAKLG LLGLANSLAI EGRKSNIHCN TIAPNAGSRM TQTVMPEDLV EALKPEYVAP LVLWLCHESC EENGGLFEVG AGWIGKLRWE RTLGAIVRQK NHPMTPEAVK ANWKKICDFE NASKPQSIQE STGSIIEVLS KIDSEGGVSA NHTSRATSTA TSGFAGAIGQ KLPPFSYAYT ELEAIMYALG VGASIKDPKD LKFIYEGSSD FSCLPTFGVI IGQKSMMGGG LAEIPGLSIN FAKVLHGEQY LELYKPLPRA GKLKCEAVVA DVLDKGSGVV IIMDVYSYSE KELICHNQFS LFLVGSGGFG GKRTSDKVKV AVAIPNRPPD AVLTDTTSLN QAALYRLSGD WNPLHIDPNF ASLAGFDKPI LHGLCTFGFS ARRVLQQFAD NDVSRFKAIK ARFAKPVYPG QTLQTEMWKE GNRIHFQTKV QETGDIVISN AYVDLAPTSG TSAKTPSEGG KLQSTFVFEE IGRRLKDIGP EVVKKVNAVF EWHITKGGNI GAKWTIDLKS GSGKVYQGPA KGAADTTIIL SDEDFMEVVL GKLDPQKAFF SGRLKARGNI MLSQKLQMIL KDYAKL | |
Greater than 90% as determined by SDS-PAGE. |