missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Peptide YY Protein
A DNA sequence encoding the Homo sapiens (Human) Peptide YY, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8738-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_004151.3 | |
Peptide YY Protein | |
Research Use Only | |
PYY | |
Recombinant Protein | |
E. coli | |
IKPEAPREDA SPEELNRYYA SLRHYLNLVT RQRY | |
Greater than 90% as determined by SDS-PAGE. |
5697 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |