missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PCK1 Protein
A DNA sequence encoding the Homo sapiens (Human) PCK1, was expressed in the hosts and tags indicated.
160.00 EUR - 2090.00 EUR
Specifications
Accession Number | NP_002582.3 |
---|---|
Gene ID (Entrez) | 5105 |
Name | PCK1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16043520
|
enQuireBio™
QP6469-EC-100UG |
100 μg |
463.00 EUR
100µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16063520
|
enQuireBio™
QP6469-EC-1MG |
1 mg |
2090.00 EUR
1mg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16053520
|
enQuireBio™
QP6469-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16073520
|
enQuireBio™
QP6469-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16083520
|
enQuireBio™
QP6469-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16093520
|
enQuireBio™
QP6469-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_002582.3 | |
PCK1 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
5105 | |
Research Use Only | |
PCK1 | |
Recombinant Protein | |
E. coli | |
MPPQLQNGLN LSAKVVQGSL DSLPQAVREF LENNAELCQP DHIHICDGSE EENGRLLGQM EEEGILRRLK KYDNCWLALT DPRDVARIES KTVIVTQEQR DTVPIPKTGL SQLGRWMSEE DFEKAFNARF PGCMKGRTMY VIPFSMGPLG SPLSKIGIEL TDSPYVVASM RIMTRMGTPV LEAVGDGEFV KCLHSVGCPL PLQKPLVNNW PCNPELTLIA HLPDRREIIS FGSGYGGNSL LGKKCFALRM ASRLAKEEGW LAEHMLILGI TNPEGEKKYL AAAFPSACGK TNLAMMNPSL PGWKVECVGD DIAWMKFDAQ GHLRAINPEN GFFGVAPGTS VKTNPNAIKT IQKNTIFTNV AETSDGGVYW EGIDEPLASG VTITSWKNKE WSSEDGEPCA HPNSRFCTPA SQCPIIDAAW ESPEGVPIEG IIFGGRRPAG VPLVYEALSW QHGVFVGAAM RSEATAAAEH KGKIIMHDPF AMRPFFGYNF GKYLAHWLSM AQHPAAKLPK IFHVNWFRKD KEGKFLWPGF GENSRVLEWM FNRIDGKAST KLTPIGYIPK EDALNLKGLG HINMMELFSI SKEFWEKEVE DIEKYLEDQV NADLPCEIER EILALKQRIS QM | |
Greater than 90% as determined by SDS-PAGE. |