missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PA28A / PSME1 Protein

A DNA sequence encoding the Homo sapiens (Human) PA28A / PSME1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6547-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16044070

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001268457.1
PA28A / PSME1 Protein
Research Use Only
PSME1
Recombinant Protein
E. coli
MAMLRVQPEA QAKVDVFRED LCTKTENLLG SYFPKKISEL DAFLKEPALN EANLSNLKAP LDIPVPDPVK EKEKEERKKQ QEKEDKDEKK KGEDEDKGPP CGPVNCNEKI VVLLQRLKPE IKDVIEQLNL VTTWLQLQIP RIEDGNNFGV AVQEKVFELM TSLHTKLEGF HTQISKYFSE RGDAVTKAAK QPHVGDYRQL VHELDEAEYR DIRLMVMEIR NAYAVLYDII LKNFEKLKKP RGETKGMIY
Greater than 80% as determined by SDS-PAGE.
5720
10 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers