missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Osteonectin / SPARC Protein

A DNA sequence encoding the Homo sapiens (Human) Osteonectin / SPARC, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8664-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16045841

  • 463.00 EUR / 100µg
Estimated Shipment: 01-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001296372.1
Osteonectin / SPARC Protein
Research Use Only
SPARC
Recombinant Protein
E. coli
APQQEALPDE TEVVEETVAE VTEVSVGANP VQVEVGEFDD GAEETEEEVV AENPCQNHHC KHGKVCELDE NNTPMCVCQD PTSCPAPIGE FEKVCSNDNK TFDSSCHFFA TKCTLEGTKK GHKLHLDYIG PCKYIPPCLD SELTEFPLRM RDWLKNVLVT LYERDEDNNL LTEKQKLRVK KIHENEKRLE AGDHPVELLA RDFEKNYNMY IFPVHWQFGQ LDQHPIDGYL SHTELAPLRA PLIPMEHCTT RFFETCDLDN DKYIALDEWA GCFGIKQKDI DKDLVI
Greater than 90% as determined by SDS-PAGE.
6678
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers