missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Osteonectin / SPARC Protein
A DNA sequence encoding the Homo sapiens (Human) Osteonectin / SPARC, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8664-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001296372.1 | |
Osteonectin / SPARC Protein | |
Research Use Only | |
SPARC | |
Recombinant Protein | |
E. coli | |
APQQEALPDE TEVVEETVAE VTEVSVGANP VQVEVGEFDD GAEETEEEVV AENPCQNHHC KHGKVCELDE NNTPMCVCQD PTSCPAPIGE FEKVCSNDNK TFDSSCHFFA TKCTLEGTKK GHKLHLDYIG PCKYIPPCLD SELTEFPLRM RDWLKNVLVT LYERDEDNNL LTEKQKLRVK KIHENEKRLE AGDHPVELLA RDFEKNYNMY IFPVHWQFGQ LDQHPIDGYL SHTELAPLRA PLIPMEHCTT RFFETCDLDN DKYIALDEWA GCFGIKQKDI DKDLVI | |
Greater than 90% as determined by SDS-PAGE. |
6678 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |