missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Origin recognition complex subunit 4 Protein
A DNA sequence encoding the Homo sapiens (Human) Origin recognition complex subunit 4, was expressed in the hosts and tags indicated.
160.00 EUR - 2090.00 EUR
Specifications
Accession Number | NP_001177808.1 |
---|---|
Gene ID (Entrez) | 5000 |
Name | Origin recognition complex subunit 4 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16003420
|
enQuireBio™
QP6453-EC-100UG |
100 μg |
463.00 EUR
100µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16023420
|
enQuireBio™
QP6453-EC-1MG |
1 mg |
2090.00 EUR
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16013420
|
enQuireBio™
QP6453-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16033420
|
enQuireBio™
QP6453-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16043420
|
enQuireBio™
QP6453-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16053420
|
enQuireBio™
QP6453-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001177808.1 | |
Origin recognition complex subunit 4 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
5000 | |
Research Use Only | |
ORC4 | |
Recombinant Protein | |
E. coli | |
MSSRKSKSNS LIHTECLSQV QRILRERFCR QSPHSNLFGV QVQYKHLSEL LKRTALHGES NSVLIIGPRG SGKTMLINHA LKELMEIEEV SENVLQVHLN GLLQINDKIA LKEITRQLNL ENVVGDKVFG SFAENLSFLL EALKKGDRTS SCPVIFILDE FDLFAHHKNQ TLLYNLFDIS QSAQTPIAVI GLTCRLDILE LLEKRVKSRF SHRQIHLMNS FGFPQYVKIF KEQLSLPAEF PDKVFAEKWN ENVQYLSEDR SVQEVLQKHF NISKNLRSLH MLLMLALNRV TASHPFMTAV DLMEASQLCS MDSKANIVHG LSVLEICLII AMKHLNDIYE EEPFNFQMVY NEFQKFVQRK AHSVYNFEKP VVMKAFEHLQ QLELIKPMER TSGNSQREYQ LMKLLLDNTQ IMNALQKYPN CPTDVRQWAT SSLSWL | |
Greater than 90% as determined by SDS-PAGE. |