missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Orexin receptor type 2 Protein

A DNA sequence encoding the Homo sapiens (Human) Orexin receptor type 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9175-ye-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16007771

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001517.2
Orexin receptor type 2 Protein
Research Use Only
HCRTR2
Recombinant Protein
Yeast
MSGTKLEDSP PCRNWSSASE LNETQEPFLN PTDYDDEEFL RYLWREYLHP KEYEWVLIAG YIIVFVVALI GNVLV
Greater than 90% as determined by SDS-PAGE.
3062
10 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers