missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Odorant-binding protein 2a Protein

A DNA sequence encoding the Homo sapiens (Human) Odorant-binding protein 2a, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8123-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16062791

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001280118.1
Odorant-binding protein 2a Protein
Research Use Only
OBP2A
Recombinant Protein
E. coli
LSFTLEEEDI TGTWYVKAMV VDKDFPEDRR PRKVSPVKVT ALGGGNLEAT FTFMREDRCI QKKILMRKTE EPGKFSAYGG RKLIYLQELP GTDDYVFYCK DQRRGGLRYM GKLVGRNPNT NLEALEEFKK LVQHKGLSEE DIFMPLQTGS CVLEH
Greater than 90% as determined by SDS-PAGE.
29991
500 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers