missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NUDT2 / Ap4A hydrolase Protein
A DNA sequence encoding the Homo sapiens (Human) NUDT2 / Ap4A hydrolase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6935-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001152.1 | |
NUDT2 / Ap4A hydrolase Protein | |
Research Use Only | |
NUDT2 | |
Recombinant Protein | |
E. coli | |
MALRACGLII FRRCLIPKVD NNAIEFLLLQ ASDGIHHWTP PKGHVEPGED DLETALRETQ EEAGIEAGQL TIIEGFKREL NYVARNKPKT VIYWLAEVKD YDVEIRLSHE HQAYRWLGLE EACQLAQFKE MKAALQEGHQ FLCSIEA | |
Greater than 80% as determined by SDS-PAGE. |
318 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |