missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NUDT2 / Ap4A hydrolase Protein

A DNA sequence encoding the Homo sapiens (Human) NUDT2 / Ap4A hydrolase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6935-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16006770

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001152.1
NUDT2 / Ap4A hydrolase Protein
Research Use Only
NUDT2
Recombinant Protein
E. coli
MALRACGLII FRRCLIPKVD NNAIEFLLLQ ASDGIHHWTP PKGHVEPGED DLETALRETQ EEAGIEAGQL TIIEGFKREL NYVARNKPKT VIYWLAEVKD YDVEIRLSHE HQAYRWLGLE EACQLAQFKE MKAALQEGHQ FLCSIEA
Greater than 80% as determined by SDS-PAGE.
318
200 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers