missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Nuclear transport factor 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Nuclear transport factor 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6442-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001308967.1 | |
Nuclear transport factor 2 Protein | |
Research Use Only | |
NUTF2 | |
Recombinant Protein | |
E. coli | |
MGDKPIWEQI GSSFIQHYYQ LFDNDRTQLG AIYIDASCLT WEGQQFQGKA AIVEKLSSLP FQKIQHSITA QDHQPTPDSC IISMVVGQLK ADEDPIMGFH QMFLLKNIND AWVCTNDMFR LALHNFG | |
Greater than 80% as determined by SDS-PAGE. |
10204 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |