missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Nuclear inhibitor of protein phosphatase 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Nuclear inhibitor of protein phosphatase 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7556-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002704.1 | |
Nuclear inhibitor of protein phosphatase 1 Protein | |
Research Use Only | |
PPP1R8 | |
Recombinant Protein | |
E. coli | |
MGGEDDELKG LLGLPEEETE LDNLTEFNTA HNKRISTLTI EEGNLDIQRP KRKRKNSRVT FSEDDEIINP EDVDPSVGRF RNMVQTAVVP VKKKRVEGPG SLGLEESGSR RMQNFAFSGG LYGGLPPTHS EAGSQPHGIH GTALIGGLPM PYPNLAPDVD LTPVVPSAVN MNPAPNPAVY NPEAVNEPKK KKYAKEAWPG KKPTPSLLIL ENGTHMASSD ACHECKSMIA AAG | |
Greater than 80% as determined by SDS-PAGE. |
5511 | |
500 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |