missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NR2F6 Protein

A DNA sequence encoding the Homo sapiens (Human) NR2F6, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6434-ma-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16013290

  • 1695.00 EUR / 100µg
Estimated Shipment: 09-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_005225.2
NR2F6 Protein
Research Use Only
NR2F6
Recombinant Protein
Mammal
MAMVTGGWGG PGGDTNGVDK AGGYPRAAED DSASPPGAAS DAEPGDEERP GLQVDCVVCG DKSSGKHYGV FTCEGCKSFF KRSIRRNLSY TCRSNRDCQI DQHHRNQCQY CRLKKCFRVG MRKEAVQRGR IPHSLPGAVA ASSGSPPGSA LAAVASGGDL FPGQPVSELI AQLLRAEPYP AAAGRFGAGG GAAGAVLGID NVCELAARLL FSTVEWARHA PFFPELPVAD QVALLRLSWS ELFVLNAAQA ALPLHTAPLL AAAGLHAAPM AAERAVAFMD QVRAFQEQVD KLGRLQVDSA EYGCLKAIAL FTPDACGLSD PAHVESLQEK AQVALTEYVR AQYPSQPQRF GRLLLRLPAL RAVPASLISQ LFFMRLVGKT PIETLIRDML LSGSTFNWPY GSGQ
Greater than 90% as determined by SDS-PAGE.
2063
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers