missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NMNAT3 Protein
A DNA sequence encoding the Homo sapiens (Human) NMNAT3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7901-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001186976.1 | |
NMNAT3 Protein | |
Research Use Only | |
NMNAT3 | |
Recombinant Protein | |
E. coli | |
MYQVIQGIIS PVNDTYGKKD LAASHHRVAM ARLALQTSDW IRVDPWESEQ AQWMETVKVL RHHHSKLLRS PPQMEGPDHG KALFSTPAAV PELKLLCGAD VLKTFQTPNL WKDAHIQEIV EKFGLVCVGR VGHDPKGYIA ESPILRMHQH NIHLAKEPVQ NEISATYIRR ALGQGQSVKY LIPDAVITYI KDHGLYTKGS TWKGKSTQST EGKTS | |
Greater than 90% as determined by SDS-PAGE. |
349565 | |
50 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |