missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Nicotinamide N-methyltransferase Protein

A DNA sequence encoding the Homo sapiens (Human) Nicotinamide N-methyltransferase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6426-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16033230

  • 2090.00 EUR / 1mg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_006160.1
Nicotinamide N-methyltransferase Protein
Research Use Only
NNMT
Recombinant Protein
E. coli
MESGFTSKDT YLSHFNPRDY LEKYYKFGSR HSAESQILKH LLKNLFKIFC LDGVKGDLLI DIGSGPTIYQ LLSACESFKE IVVTDYSDQN LQELEKWLKK EPEAFDWSPV VTYVCDLEGN RVKGPEKEEK LRQAVKQVLK CDVTQSQPLG AVPLPPADCV LSTLCLDAAC PDLPTYCRAL RNLGSLLKPG GFLVIMDALK SSYYMIGEQK FSSLPLGREA VEAAVKEAGY TIEWFEVISQ SYSSTMANNE GLFSLVARKL SRPL
Greater than 80% as determined by SDS-PAGE.
4837
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers