missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Neurofilament Protein
A DNA sequence encoding the Homo sapiens (Human) Neurofilament, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6415-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_006149.2 | |
Neurofilament Protein | |
Research Use Only | |
NEFL | |
Recombinant Protein | |
E. coli | |
SSFSYEPYYS TSYKRRYVET PRVHISSVRS GYSTARSAYS SYSAPVSSSL SVRRSYSSSS GSLMPSLENL DLSQVAAISN DLKSIRTQEK AQLQDLNDRF ASFIERVHEL EQQNKVLEAE LLVLRQKHSE PSRFRALYEQ EIRDLRLAAE DATNEKQALQ GEREGLEETL RNLQARYEEE VLSREDAEGR LMEARKGADE AALARAELEK RIDSLMDEIS FLKKVHEEEI AELQAQIQYA QISVEMDVTK PDLSAALKDI RAQYEKLAAK NMQNAEEWFK SRFTVLTESA AKNTDAVRAA KDEVSESRRL LKAKTLEIEA CRGMNEALEK QLQELEDKQN ADISAMQDTI NKLENELRTT KSEMARYLKE YQDLLNVKMA LDIEIAAYRK LLEGEETRLS FTSVGSITSG YSQSSQVFGR SAYGGLQTSS YLMSTRSFPS YYTSHVQEEQ IEVEETIEAA KAEEAKDEPP SEGEAEEEEK DKEEAEEEEA AEEEEAAKEE SEEAKEEEEG GEGEEGEETK EAEEEEKKVE GAGEEQAAKK KD | |
Greater than 83.9% as determined by SDS-PAGE. |
4747 | |
200 μg | |
Not determined. | |
Full Length of Mature Protein | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |