missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Neurofilament Protein

A DNA sequence encoding the Homo sapiens (Human) Neurofilament, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6415-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16013150

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_006149.2
Neurofilament Protein
Research Use Only
NEFL
Recombinant Protein
E. coli
SSFSYEPYYS TSYKRRYVET PRVHISSVRS GYSTARSAYS SYSAPVSSSL SVRRSYSSSS GSLMPSLENL DLSQVAAISN DLKSIRTQEK AQLQDLNDRF ASFIERVHEL EQQNKVLEAE LLVLRQKHSE PSRFRALYEQ EIRDLRLAAE DATNEKQALQ GEREGLEETL RNLQARYEEE VLSREDAEGR LMEARKGADE AALARAELEK RIDSLMDEIS FLKKVHEEEI AELQAQIQYA QISVEMDVTK PDLSAALKDI RAQYEKLAAK NMQNAEEWFK SRFTVLTESA AKNTDAVRAA KDEVSESRRL LKAKTLEIEA CRGMNEALEK QLQELEDKQN ADISAMQDTI NKLENELRTT KSEMARYLKE YQDLLNVKMA LDIEIAAYRK LLEGEETRLS FTSVGSITSG YSQSSQVFGR SAYGGLQTSS YLMSTRSFPS YYTSHVQEEQ IEVEETIEAA KAEEAKDEPP SEGEAEEEEK DKEEAEEEEA AEEEEAAKEE SEEAKEEEEG GEGEEGEETK EAEEEEKKVE GAGEEQAAKK KD
Greater than 83.9% as determined by SDS-PAGE.
4747
10 μg
Not determined.
Full Length of Mature Protein
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers