missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Nectin-4 Protein
A DNA sequence encoding the Homo sapiens (Human) Nectin-4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9847-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_112178.2 | |
Nectin-4 Protein | |
Research Use Only | |
PVRL4 | |
Recombinant Protein | |
Yeast | |
GELETSDVVT VVLGQDAKLP CFYRGDSGEQ VGQVAWARVD AGEGAQELAL LHSKYGLHVS PAYEGRVEQP PPPRNPLDGS VLLRNAVQAD EGEYECRVST FPAGSFQARL RLRVLVPPLP SLNPGPALEE GQGLTLAASC TAEGSPAPSV TWDTEVKGTT SSRSFKHSRS AAVTSEFHLV PSRSMNGQPL TCVVSHPGLL QDQRITHILH VSFLAEASVR GLEDQNLWHI GREGAMLKCL SEGQPPPSYN WTRLDGPLPS GVRVDGDTLG FPPLTTEHSG IYVCHVSNEF SSRDSQVTVD VLDPQEDSGK QVDLVSAS | |
Greater than 90% as determined by SDS-PAGE. |
81607 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |