missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Myosin regulatory light chain 2, atrial isoform Protein

A DNA sequence encoding the Homo sapiens (Human) Myosin regulatory light chain 2, atrial isoform, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8227-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16083401

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_067046.1
Myosin regulatory light chain 2, atrial isoform Protein
Research Use Only
MYL7
Recombinant Protein
E. coli
MASRKAGTRG KVAATKQAQR GSSNVFSMFE QAQIQEFKEA FSCIDQNRDG IICKADLRET YSQLGKVSVP EEELDAMLQE GKGPINFTVF LTLFGEKLNG TDPEEAILSA FRMFDPSGKG VVNKDEFKQL LLTQADKFSP AEVEQMFALT PMDLAGNIDY KSLCYIITHG DEKEE
Greater than 80% as determined by SDS-PAGE.
58498
500 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers