missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Myosin regulatory light chain 2, atrial isoform Protein
A DNA sequence encoding the Homo sapiens (Human) Myosin regulatory light chain 2, atrial isoform, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8227-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_067046.1 | |
Myosin regulatory light chain 2, atrial isoform Protein | |
Research Use Only | |
MYL7 | |
Recombinant Protein | |
E. coli | |
MASRKAGTRG KVAATKQAQR GSSNVFSMFE QAQIQEFKEA FSCIDQNRDG IICKADLRET YSQLGKVSVP EEELDAMLQE GKGPINFTVF LTLFGEKLNG TDPEEAILSA FRMFDPSGKG VVNKDEFKQL LLTQADKFSP AEVEQMFALT PMDLAGNIDY KSLCYIITHG DEKEE | |
Greater than 80% as determined by SDS-PAGE. |
58498 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |