missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Myosin regulatory lc 2, ventric/card muscle isoform Protein

A DNA sequence encoding the Homo sapiens (Human) Myosin regulatory lc 2, ventricular/cardiac muscle isoform, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8680-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16005931

  • 2090.00 EUR / 1mg
Estimated Shipment: 08-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_000423.2
Myosin regulatory lc 2, ventric/card muscle isoform Protein
Research Use Only
MYL2
Recombinant Protein
E. coli
KRAGGANSNV FSMFEQTQIQ EFKEAFTIMD QNRDGFIDKN DLRDTFAALG RVNVKNEEID EMIKEAPGPI NFTVFLTMFG EKLKGADPEE TILNAFKVFD PEGKGVLKAD YVREMLTTQA ERFSKEEVDQ MFAAFPPDVT GNLDYKNLVH IITHGEE
Greater than 90% as determined by SDS-PAGE.
4633
1 mg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers