missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Myelin transcription factor 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Myelin transcription factor 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8849-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_004526.1 | |
Myelin transcription factor 1 Protein | |
Research Use Only | |
MYT1 | |
Recombinant Protein | |
E. coli | |
GLGHISGKYA SHRSASGCPL AARRQKEGSL NGSSFSWKSL KNEGPTCPTP GCDGSGHANG SFLTHRSLSG CPRATFAGKK GKLSGDEVLS PKFKTSDVLE NDEEIKQLNQ EIRDLNESNS EMEAAMVQLQ SQISSMEKNL KNIEEENKLI EEQNEALFLE LSGLSQALIQ SLANIRLPHM EPICEQNFDA YVSTLTDMYS NQ | |
Greater than 90% as determined by SDS-PAGE. |
4661 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |