missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Myelin expression factor 2 Protein

A DNA sequence encoding the Homo sapiens (Human) Myelin expression factor 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6399-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16023040

  • 463.00 EUR / 100µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001288139.1
Myelin expression factor 2 Protein
Research Use Only
MYEF2
Recombinant Protein
E. coli
GGMNRIGGGI GFGGLEAMNS MGGFGGVGRM GELYRGAMTS SMERDFGRGD IGINQGFGDS FGRLGSAMIG GFAGRIGSSN MGPVGSGISG GMGSMNSVTG GMGMGLDRMS SSFDRMGPGI GAILERSIDM DRGFLSGPMG SGMRERIGSK GNQIFVRNLP FDLTWQKLKE KFSQCGHVMF AEIKMENGKS KGCGTVRFDS PESAE
Greater than 90% as determined by SDS-PAGE.
50804
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers