missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Muscleblind-like protein 1
A DNA sequence encoding the Homo sapiens (Human) Muscleblind-like protein 1, was expressed in the hosts and tags indicated.
160.00 EUR - 2090.00 EUR
Specifications
Accession Number | NP_001300986.1 |
---|---|
Gene ID (Entrez) | 4154 |
Name | Muscleblind-like protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16043371
|
enQuireBio™
QP8222-EC-100UG |
100 μg |
463.00 EUR
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16063371
|
enQuireBio™
QP8222-EC-1MG |
1 mg |
2090.00 EUR
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16053371
|
enQuireBio™
QP8222-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16073371
|
enQuireBio™
QP8222-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16083371
|
enQuireBio™
QP8222-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16093371
|
enQuireBio™
QP8222-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001300986.1 | |
Muscleblind-like protein 1 | |
Not determined. | |
Full Length of Isoform 5 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
4154 | |
Research Use Only | |
MBNL1 | |
Recombinant Protein | |
E. coli | |
MAVSVTPIRD TKWLTLEVCR EFQRGTCSRP DTECKFAHPS KSCQVENGRV IACFDSLKGR CSRENCKYLH PPPHLKTQLE INGRNNLIQQ KNMAMLAQQM QLANAMMPGA PLQPVPMFSV APSLATNASA AAFNPYLGPV SPSLVPAEIL PTAPMLVTGN PGVPVPAAAA AAAQKLMRTD RLEVCREYQR GNCNRGENDC RFAHPADSTM IDTNDNTVTV CMDYIKGRCS REKCKYFHPP AHLQAKIKAA QYQVNQAAAA QAAATAAAMG IPQAVLPPLP KRPALEKTNG ATAVFNTGIF QYQQALANMQ LQQHTAFLPP GSILCMTPAT SVVPMVHGAT PATVSAATTS ATSVPFAATA TANQIPIISA EHLTSHKYVT QM | |
Greater than 90% as determined by SDS-PAGE. |