missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Minpp1 Protein
A DNA sequence encoding the Homo sapiens (Human) Minpp1, was expressed in the hosts and tags indicated.
160.00 EUR - 2090.00 EUR
Specifications
Accession Number | NP_001171588.1 |
---|---|
Gene ID (Entrez) | 9562 |
Name | Minpp1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16043551
|
enQuireBio™
QP8249-EC-100UG |
100 μg |
463.00 EUR
100µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16063551
|
enQuireBio™
QP8249-EC-1MG |
1 mg |
2090.00 EUR
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16053551
|
enQuireBio™
QP8249-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16073551
|
enQuireBio™
QP8249-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16083551
|
enQuireBio™
QP8249-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16093551
|
enQuireBio™
QP8249-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001171588.1 | |
Minpp1 Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
9562 | |
Research Use Only | |
Minpp1 | |
Recombinant Protein | |
E. coli | |
SLLEPRDPVA SSLSPYFGTK TRYEDVNPVL LSGPEAPWRD PELLEGTCTP VQLVALIRHG TRYPTVKQIR KLRQLHGLLQ ARGSRDGGAS STGSRDLGAA LADWPLWYAD WMDGQLVEKG RQDMRQLALR LASLFPALFS RENYGRLRLI TSSKHRCMDS SAAFLQGLWQ HYHPGLPPPD VADMEFGPPT VNDKLMRFFD HCEKFLTEVE KNATALYHVE AFKTGPEMQN ILKKVAATLQ VPVNDLNADL IQVAFFTCSF DLAIKGVKSP WCDVFDIDDA KVLEYLNDLK QYWKRGYGYT INSRSSCTLF QDIFQHLDKA VEQKQRSQPI SSPVILQFGH AETLLPLLSL MGYFKDKEPL TAYNYKKQMH RKFRSGLIVP YASNLIFVLY HCENAKTPKE QFRVQMLLNE KVLPLAYSQE TVSFYEDLKN HYKDILQSCQ TSEECELARA NSTSDEL | |
Greater than 90% as determined by SDS-PAGE. |