missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Microtubule-associated protein 1S Protein
A DNA sequence encoding the Homo sapiens (Human) Microtubule-associated protein 1S, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8768-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001295292.1 | |
Microtubule-associated protein 1S Protein | |
Research Use Only | |
MAP1S | |
Recombinant Protein | |
E. coli | |
LSDSDPVPLA PGAADSDEDT EGFGVPRHDP LPDPLKVPPP LPDPSSICMV DPEMLPPKTA RQTENVSRTR KPLARPNSRA AAPKATPVAA AKTKGLAGGD RASRPLSARS EPSEKGGRAP LSRKSSTPKT ATRGPSGSAS SRPGVSATPP KSPVYLDLAY LPSGSSAHLV DEEFFQRVRA LCYVISGQDQ RKEEGMRAVL DALLASKQHW DRDLQVTLIP TFDSVAMHTW YAETHARHQA LGITVLGSNS MVSMQDDAFP | |
Greater than 90% as determined by SDS-PAGE. |
55201 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |