missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Metallothionein-1F Protein

A DNA sequence encoding the Homo sapiens (Human) Metallothionein-1F, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9291-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16098021

  • 1525.00 EUR / 500µg
Estimated Shipment: 02-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001288201.1
Metallothionein-1F Protein
Research Use Only
MT1F
Recombinant Protein
Yeast
MDPNCSCAAG VSCTCAGSCK CKECKCTSCK KSCCSCCPVG CSKCAQGCVC KGASEKCSC
Greater than 90% as determined by SDS-PAGE.
4494
500 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers