missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Metallothionein-1E Protein
A DNA sequence encoding the Homo sapiens (Human) Metallothionein-1E, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8895-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_783316.2 | |
Metallothionein-1E Protein | |
Research Use Only | |
MT1E | |
Recombinant Protein | |
E. coli | |
NCSCATGGSC TCAGSCKCKE CKCTSCKKSC CSCCPVGCAK CAQGCVCKGA SEKCSC | |
Greater than 90% as determined by SDS-PAGE. |
4493 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |