missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Metalloproteinase inhibitor 4 Protein
A DNA sequence encoding the Homo sapiens (Human) Metalloproteinase inhibitor 4, was expressed in the hosts and tags indicated.
160.00 EUR - 2090.00 EUR
Specifications
Accession Number | NP_003247.1 |
---|---|
Gene ID (Entrez) | 7079 |
Name | Metalloproteinase inhibitor 4 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16045351
|
enQuireBio™
QP8572-EC-100UG |
100 μg |
463.00 EUR
100µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16065351
|
enQuireBio™
QP8572-EC-1MG |
1 mg |
2090.00 EUR
1mg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16055351
|
enQuireBio™
QP8572-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16075351
|
enQuireBio™
QP8572-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16085351
|
enQuireBio™
QP8572-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16095351
|
enQuireBio™
QP8572-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_003247.1 | |
Metalloproteinase inhibitor 4 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
7079 | |
Research Use Only | |
TIMP4 | |
Recombinant Protein | |
E. coli | |
SCAPAHPQQH ICHSALVIRA KISSEKVVPA SADPADTEKM LRYEIKQIKM FKGFEKVKDV QYIYTPFDSS LCGVKLEANS QKQYLLTGQV LSDGKVFIHL CNYIEPWEDL SLVQRESLNH HYHLNCGCQI TTCYTVPCTI SAPNECLWTD WLLERKLYGY QAQHYVCMKH VDGTCSWYRG HLPLRKEFVD IVQP | |
Greater than 90% as determined by SDS-PAGE. |