missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Mediator of DNA damage checkpoint protein 1
A DNA sequence encoding the Homo sapiens (Human) Mediator of DNA damage checkpoint protein 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7613-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_055456.2 | |
Mediator of DNA damage checkpoint protein 1 | |
Research Use Only | |
MDC1 | |
Recombinant Protein | |
Yeast | |
APKVLFTGVV DARGERAVLA LGGSLAGSAA EASHLVTDRI RRTVKFLCAL GRGIPILSLD WLHQSRKAGF FLPPDEYVVT DPEQEKNFGF SLQDALSRAR ERRLLEGYEI YVTPGVQPPP PQMGEIISCC GGTYLPSMPR SYKPQRVVIT CPQDFPHCSI PLRVGLPLLS PEFLLTGVLK QEAKPEAFVL S | |
Greater than 90% as determined by SDS-PAGE. |
9656 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |