missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MBTPS1 Protein
A DNA sequence encoding the Homo sapiens (Human) MBTPS1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7551-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_003782.1 | |
MBTPS1 Protein | |
Research Use Only | |
MBTPS1 | |
Recombinant Protein | |
E. coli | |
DTGLSEKHPH FKNVKERTNW TNERTLDDGL GHGTFVAGVI ASMRECQGFA PDAELHIFRV FTNNQVSYTS WFLDAFNYAI LKKIDVLNLS IGGPDFMDHP FVDKVWELTA NNVIMVSAIG NDGPLYGTLN NPADQMDVIG VGGIDFEDNI ARFSSRGMTT WELPGGYGRM KPDIVTYGAG VRGSGVKGGC RALSGTS | |
Greater than 90% as determined by SDS-PAGE. |
8720 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |