missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MAP3K7 C-terminal-like Protein
A DNA sequence encoding the Homo sapiens (Human) MAP3K7 C-terminal-like protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7247-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001273546.1 | |
MAP3K7 C-terminal-like Protein | |
Research Use Only | |
MAP3K7CL | |
Recombinant Protein | |
E. coli | |
MISTARVPAD KPVRIAFSLN DASDDTPPED SIPLVFPELD QQLQPLPPCH DSEESMEVFK QHCQIAEEYH EVKKEITLLE QRKKELIAKL DQAEKEKVDA AELVREFEAL TEENRTLRLA QSQCVEQLEK LRIQYQKRQG SS | |
Greater than 90% as determined by SDS-PAGE. |
56911 | |
100 μg | |
Not determined. | |
Full Length of Isoform D | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |