missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Mannose-6-phosphate isomerase Protein
A DNA sequence encoding the Homo sapiens (Human) Mannose-6-phosphate isomerase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6375-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001276085.1 | |
Mannose-6-phosphate isomerase Protein | |
Research Use Only | |
MPI | |
Recombinant Protein | |
E. coli | |
AAPRVFPLSC AVQQYAWGKM GSNSEVARLL ASSDPLAQIA EDKPYAELWM GTHPRGDAKI LDNRISQKTL SQWIAENQDS LGSKVKDTFN GNLPFLFKVL SVETPLSIQA HPNKELAEKL HLQAPQHYPD ANHKPEMAIA LTPFQGLCGF RPVEEIVTFL KKVPEFQFLI GDEAATHLKQ TMSHDSMS-Q AVASSLQSCF SHLMKSEKKV VVEQLNLLVK RISQQAAAGN NMEDIFGELL LQLHQQYPGD IGCFAIYFLN LLTLKPGEAM FLEANVPHAY LKGDCVECMA CSDNTVRAGL TPKFIDVPTL CEMLSYTPSS SKDRLFLPTR SQEDPYLSIY DPPVPDFTIM KTEVPGSVTE YKVLALDSAS ILLMVQGTVI ASTPTTQTPI PLQRGGVLFI GANESVSLKL TEPKDLLIFR ACCLL | |
Greater than 80% as determined by SDS-PAGE. |
4351 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |