missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Mannan-binding lectin serine protease 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Mannan-binding lectin serine protease 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6347-ye-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001027019.1 | |
Mannan-binding lectin serine protease 1 Protein | |
Research Use Only | |
MASP1 | |
Recombinant Protein | |
Yeast | |
HTVELNNMFG QIQSPGYPDS YPSDSEVTWN ITVPDGFRIK LYFMHFNLES SYLCEYDYVK VETEDQVLAT FCGRETTDTE QTPGQEVVLS PGSFMSITFR SDFSNEERFT GFDAHYMAVD VDECKEREDE ELSCDHYCHN YIGGYYCSCR FGYILHTDNR TCRVECSDNL FTQRTGVITS PDFPNPYPKS SECLYTIELE EGFMVNLQFE DIFDIEDHPE VPCPYDYIKI KVGPKVLGPF CGEKAPEPIS TQSHSVLILF HSDNSGENRG WRLSYRAAGN ECPELQPPVH GKIEPSQAKY FFKDQVLVSC DTGYKVLKDN VEMDTFQIEC LKDGTWSNKI PTCKIVDCRA PGELEHGLIT FSTRNNLTTY KSEIKYSCQE PYYKMLNNNT GIYTCSAQGV WMNKVLGRSL PTCLPVCGLP KFSRKLMARI FNGRPAQKGT TPWIAMLSHL NGQPFCGGSL LGSSWIVTAA HCLHQSLDPE DPTLRDSDLL SPSDFKIILG KHWRLRSDEN EQHLGVKHTT LHPQYDPNTF ENDVALVELL ESPVLNAFVM PICLPEGPQQ EGAMVIVSGW GKQFLQRFPE TLMEIEIPIV DHSTCQKAYA PLKKKVTRDM ICAGEKEGGK DACAGDSGGP MVTLNRERGQ WYLVGTVSWG DDCGKKDRYG VYSYIHHNKD WIQRVTGVRN | |
Greater than 90% as determined by SDS-PAGE. |
5648 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |