missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Malate dehydrogenase, cytoplasmic Protein

A DNA sequence encoding the Homo sapiens (Human) Malate dehydrogenase, cytoplasmic, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8485-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16024881

  • 2090.00 EUR / 1mg
Estimated Shipment: 04-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001186040.1
Malate dehydrogenase, cytoplasmic Protein
Research Use Only
MDHA
Recombinant Protein
E. coli
SEPIRVLVTG AAGQIAYSLL YSIGNGSVFG KDQPIILVLL DITPMMGVLD GVLMELQDCA LPLLKDVIAT DKEDVAFKDL DVAILVGSMP RREGMERKDL LKANVKIFKS QGAALDKYAK KSVKVIVVGN PANTNCLTAS KSAPSIPKEN FSCLTRLDHN RAKAQIALKL GVTANDVKNV IIWGNHSSTQ YPDVNHAKVK LQGKEVGVYE ALKDDSWLKG EFVTTVQQRG AAVIKARKLS SAMSAAKAIC DHVRDIWFGT PEGEFVSMGV ISDGNSYGVP DDLLYSFPVV IKNKTWKFVE GLPINDFSRE KMDLTAKELT EEKESAFEFL SS
Greater than 90% as determined by SDS-PAGE.
4190
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers